missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Goat IgG Fc Cross-Adsorbed Secondary Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen A16152
Additional Details : Weight : 0.01000kg
Description
Goat IgG Fc Cross-Adsorbed Polyclonal antibody specifically detects Goat IgG Fc Cross-Adsorbed in Goat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry.Specifications
Goat IgG Fc Cross-Adsorbed | |
Polyclonal | |
Biotin | |
Rabbit | |
Affinity chromatography | |
RUO | |
Goat | |
Liquid |
ELISA, Flow Cytometry, Immunocytochemistry | |
1.5 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.2 | |
Gamma Immunoglobin Fc region | |
1 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |