missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSF Monoclonal Antibody (S1390), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA126098
Code : LD
Additional Details : Weight : 0.01000kg
Description
MA1-26098 detects GCSF Receptor from human samples.
G-CSF Monoclonal antibody specifically detects G-CSF in Human samples. It is validated for ELISA, Western Blot, Western Blot (Agglutination).Specifications
G-CSF | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P09919 | |
Csf3 | |
Purified, recombinant human MBP-GCSFR fusion protein. | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Western Blot, Western Blot (Agglutination) | |
S1390 | |
Unconjugated | |
Csf3 | |
C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF 3; CSF3; CSF3OS; Csfg; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; granulocyte-colony stimulating factor; gsf3; lenograstim; MGC45931; MGI-IG; Pluripoietin; RP23-395E10.4 | |
Mouse | |
Protein G | |
RUO | |
1440 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |