Learn More
Granzyme H Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA547129
Code : NEW
Additional Details : Weight : 0.00010kg
Description
This antibody detects both the pro and active forms of human Granzyme H in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human Granzyme B is observed and approximately 5% cross-reactivity with recombinant mouse (rm) Granzyme C, rmGranzyme D, and rmGranzyme G is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.01 EU per 1 μg of the antibody by the LAL method.
Granzyme H Polyclonal antibody specifically detects Granzyme H in Human samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
Granzyme H | |
Polyclonal | |
Unconjugated | |
GZMH | |
Cathepsin G-like 2; cathepsin G-like 2, protein h-CCPX; CCP-X; CGL2; CGL-2; CSP-C; CTLA1; CTSGL2; Cytotoxic serine protease C; Cytotoxic T-lymphocyte proteinase; cytotoxic T-lymphocyte-associated serine esterase 1; cytotoxin serine protease-C; granzyme H; granzyme H (cathepsin G-like 2, protein h-CCPX); GZMH | |
Goat | |
Antigen affinity chromatography | |
RUO | |
2999 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P20718 | |
GZMH | |
Mouse myeloma cell line NS0-derived recombinant human Granzyme H Glu19-Leu246 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |