Learn More
HA Tag Monoclonal Antibody (2-2.2.14), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 26183
252.70 EUR valid until 2024-04-30
Use promo code "21741" to get your promotional price.
Code : A0
Additional Details : Weight : 0.45360kg
Description
26183 is specific for the HA peptide YPYDVPDYA. The antibody has been used successfully in Western blot and immunoprecipitation applications. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
HA Tag Monoclonal antibody specifically detects HA Tag in Tag samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
HA Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
2-2.2.14 | |
Unconjugated | |
HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
HA peptide YPYDVPDYA derivitized to ovalbumin. | |
100 μg | |
Primary | |
Antibody | |
IgG1 |