missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLCS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529050
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: K562. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HLCS Polyclonal antibody specifically detects HLCS in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
HLCS | |
Polyclonal | |
Unconjugated | |
HLCS | |
410I21.SP6; biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase; Biotin apo-protein ligase; biotin--[acetyl-CoA-carboxylase] ligase; Biotin--[methylcrotonoyl-CoA-carboxylase] ligase; biotin--[methylmalonyl-CoA-carboxytransferase] ligase; Biotin--[propionyl-CoA-carboxylase [ATP-hydrolyzing]] ligase; Biotin--protein ligase; D16Jhu34; HCS; Hlcs; Holocarboxylase synthetase; holocarboxylase synthetase (biotin- [propriony-Coenzyme A-carboxylase (ATP-hydrolysing)] ligase); holocarboxylase synthetase (biotin-(proprionyl-CoA-carboxylase (ATP-hydrolysing)) ligase); holocarboxylase synthetase (biotin-(proprionyl-Coenzyme A-carboxylase (ATP-hydrolysing)) ligase); RGD1565360 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3141 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
P50747 | |
HLCS | |
Recombinant fragment corresponding to a region within amino acids 20 and 300 of Human HCS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |