Learn More
HIF1A Monoclonal Antibody (H1alpha 67-7), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA116517
Code : LD
Additional Details : Weight : 0.01000kg
Description
In Western blot, multiple bands may be seen at 120 kDa representing post-translational modification of HIF-1 alpha. Nuclear extracts are recommended. Suggested positive control: Cos7 CoCl2-treated nuclear extract.
HIF1A Monoclonal antibody specifically detects HIF1A in Human, Non-human Primate samples. It is validated for Western Blot.Specifications
HIF1A | |
Monoclonal | |
1.5 mg/mL | |
PBS with 0.05% sodium azide | |
Q16665 | |
HIF1A | |
Fusion protein containing residues 432-528 of human HIF-1 alpha. | |
100 μL | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG2b |
Western Blot | |
H1alpha 67-7 | |
Unconjugated | |
HIF1A | |
AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L home | |
Mouse | |
Protein G | |
RUO | |
3091 | |
4° C, do not freeze | |
Liquid |