Learn More
HP1BP3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544637
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: SEESVSTVEE QENETPPATS SEAEQPKGEP ENEEKEENKS SEETKKDEKD Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
HP1BP3 Polyclonal antibody specifically detects HP1BP3 in Human samples. It is validated for Western Blot.Specifications
HP1BP3 | |
Polyclonal | |
Unconjugated | |
HP1BP3 | |
heterochromatin protein 1 binding protein 3; heterochromatin protein 1, binding protein 3; heterochromatin protein 1-binding protein 3; Hp1bp3; HP1BP74; HP1-BP74; Protein HP1-BP74 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
50809 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q5SSJ5 | |
HP1BP3 | |
Synthetic peptide directed towards the N-terminal of human HP1BP3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |