Learn More
Hyaluronic acid Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA185561
Code : LD
Additional Details : Weight : 0.01000kg
Description
For direct ELISA, coat wells with 1% glutaraldehyde for 5 min at room temperature, followed by 3xPBS wash steps prior to coating with hyaluronic acid. Sheep anti hyaluronic acid antibody recognizes hyaluronic acid, a mucopolysaccharide which is an integral part of the gel-like substance of animal connective tissue that serves as a lubricating and protective agent.
Hyaluronic acid Polyclonal antibody specifically detects Hyaluronic acid in Chemical samples. It is validated for ELISA.Specifications
Hyaluronic acid | |
Polyclonal | |
Unconjugated | |
C28H44N2O23 | |
Hyaluronic acid from human umbilical cord. | |
1 mL | |
Primary | |
Antibody | |
IgG |
ELISA | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Sheep | |
Protein G | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Chemical |