Learn More
IL-12 p70 Polyclonal Antibody, Biotin, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA184321
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 0.5 mL PBS containing 0.1% BSA. Prior to reconstitution, store at 4oC. After reconstitution, store undiluted at -20oC, avoiding freeze/thaw cycles. This antibody may be used in Western Blotting applications under either reducing or non-reducing conditions. Goat anti Mouse Interleukin-12 antibody recognizes mouse interleukin-12 (IL-12).
IL-12 p70 Polyclonal antibody specifically detects IL-12 p70 in Mouse samples. It is validated for ELISA, Western Blot.Specifications
IL-12 p70 | |
Polyclonal | |
Biotin | |
IL12A | |
20C2; CLMF; CLMF p35; CLMF p40; CLMF2; cytokine; cytotoxic lymphocyte maturation factor 1, p35; cytotoxic lymphocyte maturation factor 35 kDa subunit; Cytotoxic lymphocyte maturation factor 40 kDa subunit; il 12; IL 12p70; Il12; IL-12; Il-12 p35; IL-12 p75; IL-12 subunit p35; IL-12 subunit p40; IL-12, subunit p35; IL12, subunit p40; Il-12; cytokine; IL12A; IL-12A; IL12A/B; IL12AB; IL12B; Il-12b; IL12P35; IL-12p35; Il12p40; Il-12p40; IL35 subunit; ILN; IMD28; IMD29; Interleukin; interleukin 12 35 kDa subunit; interleukin 12 35kDa subunit; interleukin 12 40 kDa subunit; interleukin 12 40kDa subunit; interleukin 12 p35 subunit; interleukin 12 p40 subunit precursor; interleukin 12 polypeptide 35; interleukin 12, p35; interleukin 12, p40; interleukin 12A; interleukin 12A (natural killer cell stimulatory factor 1, cytotoxic lymphocyte maturation factor 1, p35); interleukin 12B; interleukin 12B (natural killer cell stimulatory factor 2, cytotoxic lymphocyte maturation factor 2, p40); interleu | |
Goat | |
Affinity chromatography | |
RUO | |
16159, 16160 | |
-20°C | |
Lyophilized |
ELISA, Western Blot | |
0.1 mg/mL | |
PBS with no preservative | |
P43431, P43432 | |
IL12A, Il12b | |
Recombinant mouse IL-12 | |
50 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |