Learn More
ILK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528571
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (81%), Xenopus laevis (84%), Chicken (86%), Sheep (97%), Rhesus Monkey (100%), Bovine (98%), Guinea pig (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ILK Polyclonal antibody specifically detects ILK in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
ILK | |
Polyclonal | |
Unconjugated | |
ILK | |
59 kDa serine/threonine-protein kinase; AA511515; epididymis secretory protein Li 28; ESTM24; HEL-S-28; ILK; ILK1; ILK-1; ILK2; ILK-2; integrin binding protein kinase; Integrin linked ILK; integrin linked kinase; integrin-linked kinase; integrin-linked kinase-2; integrin-linked protein kinase; loc; lost-contact; main squeeze; msq; P59; p59ILK; zgc:65842; zilk | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3611 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q13418 | |
ILK | |
Recombinant fragment corresponding to a region within amino acids 1 and 230 of Human ILK | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |