missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV3.3 (KCNC3) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541024
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: YAERIGADPD DILGSNHTYF KNIPIGFWWA VVTMTTLGYG DMYPKTWSGM Sequence homology: Cow: 93%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%
KV3.3 (KCNC3) Polyclonal antibody specifically detects KV3.3 (KCNC3) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
KV3.3 (KCNC3) | |
Polyclonal | |
Unconjugated | |
KCNC3 | |
KCNC3; Kcr2-3; KShIIID; KV3.3; LOW QUALITY PROTEIN: potassium voltage-gated channel subfamily C member 3; potassium channel, voltage gated Shaw related subfamily C, member 3; potassium channel, voltage gated Shaw-related subfamily C, member 3; potassium voltage gated channel, Shaw-related subfamily, member 3; potassium voltage-gated channel subfamily C member 3; potassium voltage-gated channel, Shaw-related subfamily, member 3; SCA13; Shaw type potassium channel Kv3.3; Shaw-related voltage-gated potassium channel protein 3; voltage-gated potassium channel protein KV3.3; voltage-gated potassium channel subunit Kv3.3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3748 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14003 | |
KCNC3 | |
Synthetic peptide directed towards the middle region of human KCNC3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |