missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LC3B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA535195
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
LC3B Polyclonal antibody specifically detects LC3B in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
LC3B | |
Polyclonal | |
Unconjugated | |
MAP1LC3B2 | |
1010001C15Rik; Atg8; ATG8E; ATG8F; ATG8G; Autophagy-related protein LC3 B; autophagy-related ubiquitin-like modifier LC3 B; LC3; LC3 beta; LC3A; LC3B; light chain 3; MAP1 light chain 3-like protein 2; MAP1A/1B light chain 3 B; MAP1A/1BLC3; MAP1A/MAP1B LC3 A; MAP1A/MAP1B LC3 B; MAP1A/MAP1B light chain 3 B; Map1alc3; MAP1BLC3; MAP1LC3; MAP1LC3A; MAP1LC3B; MAP1LC3B2; MAP1LC3B-a; microtubule associated protein 1 light chain 3 beta; microtubule associated protein 1 light chain 3 beta 2; microtubule-associated protein 1 light chain 3; microtubule-associated protein 1 light chain 3 beta; microtubule-associated protein 1 light chain 3 beta 2; microtubule-associated proteins 1A/1B light chain 3; microtubule-associated proteins 1A/1B light chain 3 beta 2; Microtubule-associated proteins 1A/1B light chain 3B; Microtubule-associated proteins 1A/1B light chain 3B-like; Microtubule-associated proteins 1A/1B light chain 3B-like protein; Mpl3; RP11-346K17.1; wu:fb60g11; zbs559; zgc:56434 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
100359928, 643246, 64862, 67443, 81631 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.50 mg/mL | |
PBS with 0.09% sodium azide | |
A6NCE7, Q62625, Q9CQV6, Q9GZQ8 | |
MAP1LC3B, MAP1LC3B2 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human LC3 | |
200 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |