missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LSD1 Monoclonal Antibody (1B2E5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 413300
Additional Details : Weight : 0.50000kg
Description
LSD1 Monoclonal antibody specifically detects LSD1 in Bovine, Canine, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
LSD1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
O60341, Q6ZQ88 | |
KDM1A | |
Recombinant protein derived from the internal region of human LSD1 protein | |
100 μg | |
Primary | |
Bovine, Canine, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1B2E5 | |
Unconjugated | |
KDM1A | |
1810043O07Rik; AA408884; amine oxidase (flavin containing) domain 2; AOF2; BHC110; BRAF35-HDAC complex protein BHC110; CPRF; D4Ertd478e; FAD-binding protein BRAF35-HDAC complex, 110 kDa subunit; flavin-containing amine oxidase domain-containing protein 2; KDM1; KDM1A; KIAA0601; Lsd1; lysine (K)-specific demethylase 1; lysine (K)-specific demethylase 1A; lysine demethylase 1A; lysine-specific demethylase 1; lysine-specific histone demethylase 1; lysine-specific histone demethylase 1A; mKIAA0601; neuroprotective protein 3; RGD1562975; RP1-184J9.1; wu:fb82e06; wu:fc51a02; wu:fj36a05; wu:fj66b06 | |
Mouse | |
Protein A | |
RUO | |
23028, 478193, 500569, 532997, 99982 | |
-20°C | |
Liquid |