Learn More
Luteinizing Hormone Monoclonal Antibody (5304), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514660
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-14660 targets Luteinizing Hormone in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MA5-14660 immunogen is human Luteinizing Hormone. MA5-14660 detects Luteinizing Hormone which has a predicted molecular weight of approximately 13 kDa. Product MA514660 is a smaller package size of MBL0110 (formerly sold as a Seradyn product).
Luteinizing Hormone Monoclonal antibody specifically detects Luteinizing Hormone in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmune Assays (RIA).Specifications
Luteinizing Hormone | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P01215, P01229 | |
Cga, LHB | |
Human Luteinizing Hormone | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
5304 | |
Unconjugated | |
Cga | |
aGSU; alphaGSU; alpha-GSU; alphaSU; Anterior pituitary glycoprotein hormones common subunit alpha; CGA; Cga1; CG-alpha; CGB4; Choriogonadotropin alpha chain; chorionic gonadotrophin subunit alpha; chorionic gonadotropin, alpha polypeptide; follicle-stimulating hormone alpha chain; follicle-stimulating hormone alpha subunit; follitropin; follitropin alpha chain; FSHA; FSH-alpha; FSH-B; FSH-beta; glycoprotein hormones alpha chain; glycoprotein hormones, alpha polypeptide; glycoprotein hormones, alpha subunit; GPHa; GPHA1; GPHalpha; HCG; HH23; hLHB; interstitial cell stimulating hormone, beta chain; interstitial cell-stimulating hormone; leutenising hormone beta; leutropin; LH; LH alpha 1-1; LH alpha 1-2; LH alpha 1-3; LH alpha 3; LH beta-1; LH beta-2; LH beta-3; LHA; LHB; LH-B; LHbeta; LSH-alpha; LSH-B; LSH-beta; Luteinizing hormone (lutropin) subunit beta; luteinizing hormone alpha chain; luteinizing hormone beta; luteinizing hormone beta polypeptide; luteinizing hormone beta subunit; l | |
Mouse | |
Protein A | |
RUO | |
1081, 3972 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |