missing translation for 'onlineSavingsMsg'
Learn More
Learn More
M13 Phage coat protein Monoclonal Antibody (E1), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA112898
Code : LD
Additional Details : Weight : 0.01000kg
Description
M13 Phage coat protein Monoclonal antibody specifically detects M13 Phage coat protein in Virus samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry, Western Blot.Specifications
M13 Phage coat protein | |
Monoclonal | |
1 mg/mL | |
PBS with 0.08% sodium azide | |
M13 phage coat proteins. | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
E1 | |
Biotin | |
Mouse | |
Protein A/G | |
RUO | |
Virus | |
Antibody | |
IgG3 |