missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIF Monoclonal Antibody (2Ar3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA120881
Code : LD
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
MIF Monoclonal antibody specifically detects MIF in Human samples. It is validated for Immunocytochemistry, Western Blot.Specifications
MIF | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and no preservative | |
P14174 | |
Mif | |
E. coli-derived recombinant full length protein (Human). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
2Ar3 | |
Unconjugated | |
Mif | |
cytokine; delayed early response protein 6; DER6; GIF; GLIF; glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; HGNC:7097; L-dopachrome isomerase; L-dopachrome tautomerase; macrophage migration inhibitory factor; macrophage migration inhibitory factor (glycosylation-inhibiting factor); MGC107654; Mif; MMIF; phenylpyruvate tautomerase | |
Mouse | |
Protein G | |
RUO | |
4282 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |