Learn More
Moesin Monoclonal Antibody (38/87), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514043
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-14043 targets Moesin in IF, IHC (P), IP, and WB applications and shows reactivity with Bovine, Human, and Rat samples. The MA5-14043 immunogen is moesin purified from cow uterus.
Moesin Monoclonal antibody specifically detects Moesin in Bovine, Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Moesin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
O35763, P26038, Q2HJ49 | |
MSN | |
Moesin purified from cow uterus | |
500 μL | |
Primary | |
Bovine, Human, Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
38/87 | |
Unconjugated | |
MSN | |
C78546; epididymis luminal protein 70; HEL70; Membrane-organizing extension spike protein; MGC137877 protein; moesin; MSN | |
Mouse | |
Protein G | |
RUO | |
4478, 540426, 81521 | |
4° C | |
Liquid |