Learn More
anti-Mre11, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
POA
Description
Description
Mre11 Polyclonal specifically detects Mre11 in Human, Mouse, Rat, Chicken, Hamster samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Proximity Ligation Assay, Knockout Validated, Knockdown Validated.