Learn More
eNOS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1037
Code : LE
Additional Details : Weight : 0.10000kg
Description
PA1-037 detects endothelial nitric oxide synthase (eNOS) from bovine, canine, human, mouse, porcine and rat tissues as well as recombinant protein. This antibody does not detect human iNOS or rat nNOS. PA1-037 has been successfully used in Western blot and immunohistochemistry (paraffin and frozen) procedures. By Western blot, this antibody detects an ∽140 kDa protein representing eNOS from human endothelial cells. By Western blot PA1-037 also detects eNOS in rat lung, brain, and canine cardiac microsomes. Immunohistochemical staining of eNOS in human vascular endothelial cells with PA1-037 results in prominent staining of the epithelial cells. The PA1-037 immunogen is a synthetic peptide corresponding to residues S(1179) L Q E R Q L R G A V P W A F D(1194) of human eNOS.
eNOS Polyclonal antibody specifically detects eNOS in Bovine, Canine, Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
eNOS | |
Polyclonal | |
Unconjugated | |
NOS3 | |
2310065A03Rik; cNOS; Constitutive NOS; Ecnos; EC-NOS; endothelial nitric oxide synthase; endothelial nitric oxide synthase 3; endothelial nitric oxide synthase NOS3; endothelial NOS; eNos; e-NOS; nitric oxide synthase; nitric oxide synthase 3; nitric oxide synthase 3 (endothelial cell); nitric oxide synthase 3 transcript variant eNOS-delta20; nitric oxide synthase 3 transcript variant eNOS-delta20-21; nitric oxide synthase 3 transcript variant eNOS-delta21; nitric oxide synthase 3, endothelial cell; nitric oxide synthase, endothelial; NOS; NOS type III; Nos3; Nos-3; NOSIII; OTTHUMP00000213183 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
18127, 24600, 287024, 397557, 403784, 4846 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P29473, P29474, P70313, Q28969, Q62600, Q9TUX8 | |
NOS3 | |
Synthetic peptide corresponding to residues S(1179) L Q E R Q L R G A V P W A F D(1194) of human eNOS. | |
100 μL | |
Primary | |
Bovine, Canine, Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |