Learn More
OCT4 Monoclonal Antibody (9B7), HRP, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1104HRP
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA1-104-HRP detects Oct4/Pou5F1 in human embryonic carcinoma cells. By Western blot, this antibody detects a band at 40 kDa in human embryonal carcinoma cells, but not in negative control cells.
OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Western Blot.Specifications
OCT4 | |
Monoclonal | |
1 mg/mL | |
proprietary buffer with proprietary stabilizer and 0.07% Kathon | |
Q01860 | |
POU5F1 | |
Full length human recombinant protein of human POU5F1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
9B7 | |
HRP | |
POU5F1 | |
2-Oct; 3-Oct; 4-Oct; cb197; chunp6868; DADB-104B20.2; etID49452.21; gp9; gp-9; HGNC:9221; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; MGC22487; NF-A3; Oct2; Oct-2; Oct2a; Oct2b; OCT3; Oct-3; Oct3/4; Oct-3/4; OCT4; Oct-4; Octamer-binding protein 2; octamer-binding protein 3; Octamer-binding protein 4; octamer-binding transcription factor 2; Octamer-binding transcription factor 3; OTF2; OTF-2; OTF3; Otf-3; Otf3g; Otf3-rs7; Otf4; Otf-4; OTTHUMP00000221150; OTTHUMP00000221151; POU class 2 homeobox 2; POU class 5 homeobox 1; POU class 5 homeobox 1 transcript variant OCT4B1; POU class 5 homeobox 1 transcript variant OCT4B2; POU class 5 homeobox 1 transcript variant OCT4B3; POU class 5 homeobox 1 transcript variant OCT4B4; POU class 5 homeobox 1 transcript variant OCT4B5; POU class 5 homeobox 1 transcript variant OCT4B6; POU domain class 5 transcription factor 1; POU domain gene 2; POU domain protein 2; POU domain transcription factor OCT4; POU domain, | |
Mouse | |
Protein A | |
RUO | |
5460 | |
4° C, do not freeze | |
Liquid |