missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEBP1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518032
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine and mouse based on sequence homology.
PEBP1 Polyclonal antibody specifically detects PEBP1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
PEBP1 | |
Polyclonal | |
Unconjugated | |
PEBP1 | |
23 kDa morphine-binding protein; Basic cytosolic 21 kDa protein; epididymis luminal protein 210; epididymis secretory protein Li 34; epididymis secretory protein Li 96; HCNP; HCNPpp; HEL-210; HEL-S-34; HEL-S-96; Hippocampal cholinergic neurostimulating peptide; KIAA0299; LDGF; MDGF; MOCA; neuropolypeptide h3; P23K; PBP; Pbp1; Pbpr; PEBP; PEBP1; PEBP-1; phosphatidylethanolamine binding protein; phosphatidylethanolamine binding protein 1; phosphatidylethanolamine-binding protein; phosphatidylethanolamine-binding protein 1; PPBP; prostatic binding protein; Prostatic-binding protein; Raf kinase inhibitor protein; Raf kinase inhibitory protein; Raf-1 inhibitor protein; Raf-1 kinase inhibitor protein; RKIP; similar to phosphatidylethanolamine binding protein; zgc:56033; zgc:76942 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5037 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P30086 | |
PEBP1 | |
Synthetic peptide sequence (DYVPKLYEQLSGK) corresponding to the C-terminus amino acids of PEBP1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |