Learn More
Phospho-CREB (Ser133) Monoclonal Antibody (F.959.4), Invitrogen™
Rabbit Monoclonal Antibody
Brand: Invitrogen MA511192
Code : NEW
Additional Details : Weight : 0.10000kg
Description
Antibodies to this protein (and modification) were previously sold as part of a Thermo Scientific Cellomics High Content Screening Kit. This replacement antibody is now recommended for researchers who need an antibody for high content cell based assays. It has been thoroughly tested and validated for cellular immunofluorescence (IF) applications. Further optimization including the selection of the most appropriate fluorescent Dylight conjugated secondary antibody may have to be performed for your high content assay. It is not recommended to aliquot this antibody.
Phospho-CREB (Ser133) Monoclonal antibody specifically detects Phospho-CREB (Ser133) in Human, Mouse, Rat samples. It is validated for ChIP Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-CREB (Ser133) | |
Monoclonal | |
58 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
P15337, P16220, Q01147 | |
CREB1 | |
Synthetic phosphopeptide corresponding to residues surrounding pSer133 of human CREB | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ChIP Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
F.959.4 | |
Unconjugated | |
CREB1 | |
2310001E10Rik; 3526402H21Rik; active transcription factor CREB; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CREB; CREB1; Creb-1; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; htCREB; MGC9284; OTTHUMP00000206661; OTTHUMP00000206662; OTTHUMP00000206667; transactivator protein; Y protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
12912, 1385, 81646 | |
-20°C | |
Liquid |