missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-CrkL (Tyr207) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA517741
Code : LE
Additional Details : Weight : 0.01000kg
Description
It is not recommended to aliquot this antibody.
Phospho-CrkL (Tyr207) Polyclonal antibody specifically detects Phospho-CrkL (Tyr207) in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
Phospho-CrkL (Tyr207) | |
Polyclonal | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P46109, P47941, Q5U2U2 | |
CRKL | |
Synthetic phosphopeptide corresponding to residues surrounding pTyr207 of human CrkL | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
CRKL | |
1110025F07Rik; AA589403; AI325100; CRK like proto-oncogene, adaptor protein; CRKL; crkl {ECO:0000312; crk-like protein; crk-like protein {ECO:0000250; Crkol; EMBL:AAH85865.1, ECO:0000312; OTTHUMP00000198539; RGD:1308531}; UniProtKB:P46109}; v-crk avian sarcoma virus CT10 oncogene homolog-like; v-crk sarcoma virus CT10 oncogene homolog (avian)-like; v-crk sarcoma virus CT10 oncogene homolog-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12929, 1399, 287942 | |
-20°C | |
Liquid |