Learn More
Phospho-p38 MAPK (Thr180, Tyr182) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA184807
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Predicted to react with bovine, chicken, dog, monkey, mouse and zebrafish based on sequence homology. For the detection of phosphoproteins, serine phosphatase inhibitors such as 10mM NaF should be added to the sample buffer. Milk or other casein-based blocking solutions are not recommended as casein is a phosphoprotein and its use can result in high background. In Western blot, this antibody detects a band at ∽39kDa in Western blots of anisomycin C-6 glioma cell lysates. Rabbit anti Rat p38 MAPK (pThr180/pTyr182) antibody recognizes mitogen-activated protein kinase p38 (p38 MAPK), also known as mitogen-activated protein kinase 14 (MAPK 14), when phosphorylated at threonine 180 and tyrosine 182.
Phospho-p38 MAPK (Thr180, Tyr182) Polyclonal antibody specifically detects Phospho-p38 MAPK (Thr180, Tyr182) in Human, Rat samples. It is validated for Western Blot.Specifications
Phospho-p38 MAPK (Thr180, Tyr182) | |
Polyclonal | |
0.01M HEPES with 0.01% BSA, 50% glycerol and 0.09% sodium azide; pH 7.5 | |
P70618, Q16539 | |
MAPK14 | |
Synthetic phosphopeptide corresponding to an amino acid sequence within p38 MAPK which includes phosphorylated Thr180 and Tyr182. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
MAPK14 | |
186F5S; anon-sts23; BG:DS00797.3; CG7393; CG7393-PA; CRK1; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; Csbp2; CSPB1; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Dm p38b; Dmel\CG7393; Dmel_CG7393; Dmp38b; Dp38; D-p38; D-p38 MAPK; Dp38b; D-p38b; EC 2.7.11.24; ERK1; ERT2; ESTS:186F5S; etID18993.8; Exip; fk28c03; hm:zeh1243; Hog; I79_016605; LD24658p; MAP 14; MAP kinase 1; MAP kinase 11; MAP kinase 14; MAP kinase 14A; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38; MAP kinase p38 alpha; MAP kinase p38 beta; MAP kinase p38a; MAP kinase p38alpha; MAP kinase p38b; MAPK; MAPK 11; MAPK 14; MAPK 14A; MAPK1 antibody; Mapk11; Mapk14; mapk14a; MAX-interacting protein 2; mitogen activated protein kinase 11; mitogen activated protein kinase 14; mitogen-activated protein kinase 11; Mitogen | |
Rabbit | |
Affinity chromatography | |
RUO | |
1432, 81649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |