missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-RSK1 (Ser221) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 44924G
Additional Details : Weight : 0.50000kg
Description
Phospho-RSK1 (Ser221) Polyclonal antibody specifically detects Phospho-RSK1 (Ser221) in Chicken, Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Phospho-RSK1 (Ser221) | |
Polyclonal | |
Dulbecco's PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3 | |
P18652, P18653, Q15418 | |
RPS6KA1 | |
The antiserum was produced against a chemically synthesized phosphopeptide derived from a region of human RSK1 that contains serine 221. The sequence is conserved in mouse and rat. | |
100 μL | |
Primary | |
Chicken, Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
RPS6KA1 | |
90 kDa ribosomal protein S6 kinase 1; dJ590P13.1 (ribosomal protein S6 kinase, 90kD, polypeptide 1); hu-1; MAP kinase-activated protein kinase 1; MAP kinase-activated protein kinase 1a; MAPK-activated protein kinase 1; MAPK-activated protein kinase 1a; MAPKAP kinase 1; MAPKAP kinase 1a; MAPKAPK-1; mapkapk1a; MAPKAPK-1a; p90Rsk; P90-RSK; p90-RSK 1; p90Rsk1; p90-Rsk1; p90S6K; pp90RSK1; ribosomal protein S6 kinase 2 alpha; ribosomal protein S6 kinase 2 beta; ribosomal protein S6 kinase a, polypeptide 1; ribosomal protein S6 kinase A1; ribosomal protein S6 kinase A1 L homeolog; ribosomal protein S6 kinase alpha 1; Ribosomal protein S6 kinase alpha-1; Ribosomal protein S6 kinase II alpha; ribosomal protein S6 kinase II beta; ribosomal protein S6 kinase polypeptide 1; ribosomal protein S6 kinase, 90kD, polypeptide 1; Ribosomal protein S6 kinase, 90kD, 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1; ribosomal protein S6 kinase, 90kDa, polypeptide 1 L homeolog; ribosomal protein S6 kinas | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20111, 386579, 6195 | |
-20°C | |
Liquid |