missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIWIL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534341
Code : NEW
Additional Details : Weight : 0.01000kg
Description
A suggested positive control is human brain tissue lysate. PA5-34341 can be used with blocking peptide PEP-1384.
PIWIL2 Polyclonal antibody specifically detects PIWIL2 in Human samples. It is validated for Immunohistochemistry.Specifications
PIWIL2 | |
Polyclonal | |
Unconjugated | |
Piwil2 | |
80kDa PIWIL2 short isoform; cancer/testis antigen 80; CT80; HILI; mili; Miwi like; Miwi likw; piwi like homolog 2; piwi like RNA-mediated gene silencing 2; PIWIL1L; Piwil2; PIWI-L2; piwil2-like protein; piwi-like 2; piwi-like homolog 2; piwi-like protein 2; piwi-like RNA-mediated gene silencing 2; testicular tissue protein Li 141 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55124 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8TC59 | |
Piwil2 | |
A 14 amino acid synthetic peptide near the center of human PIWI-L2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |