missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G2A Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA547672
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
PLA2G2A Polyclonal antibody specifically detects PLA2G2A in Mouse samples. It is validated for Immunohistochemistry (Frozen), Western Blot.Specifications
PLA2G2A | |
Polyclonal | |
Unconjugated | |
PLA2G2A | |
EC 3.1.1.4; EF; Enhancing factor; eted enzyme type IIA phospholipase A2; GIIC sPLA2; Group IIA phospholipase A2; modifier of Min1; Mom1; non-pancreatic secreted type II phospholipase A2; non-pancreatic secretory phospholipase A2; NPS-PLA2; phosphatidylcholine 2-acylhydrolase 2A; phospholipase A2 group IIA; phospholipase A2, group IIA (platelets, synovial fluid); phospholipase A2, membrane associated; Pla2; PLA2B; PLA2BPLA2; PLA2G2A; PLA2L; PLA2LPLA2S; PLA2S; PLAS1; platelet phospholipase A2; RASF-A; secretory group II phospholipase A2; sPLA2; sPla2-IIA | |
Sheep | |
Antigen affinity chromatography | |
RUO | |
18780 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P31482 | |
PLA2G2A | |
Sf 21-derived recombinant mouse PLA2G2A Asn22-Cys146 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |