Learn More
Prolactin Monoclonal Antibody (P4C4A12), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MIP0201
Code : LD
Additional Details : Weight : 0.10000kg
Description
MIP0201 targets Prolactin in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MIP0201 immunogen is human Prolactin. MIP0201 detects Prolactin which has a predicted molecular weight of approximately 25 kDa. MIP0201 was formerly sold as a Seradyn product.
Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for ELISA, Immunoprecipitation, Radioimmune Assays (RIA).Specifications
Prolactin | |
Monoclonal | |
5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
P01236 | |
PRL | |
Human Prolactin | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
P4C4A12 | |
Unconjugated | |
PRL | |
AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
Mouse | |
Protein A | |
RUO | |
5617 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |