missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTH1R Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534131
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Monkey, Marmoset (100%) Gorilla, Gibbon, Bovine, Pig (95%) Orangutan, Elephant, Dog (90%).
PTH1R Polyclonal antibody specifically detects PTH1R in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).Specifications
PTH1R | |
Polyclonal | |
Unconjugated | |
Pth1r | |
MGC138426; MGC138452; Parathormone; Parathyrin; parathyroid hormone 1 receptor; parathyroid hormone receptor; parathyroid hormone receptor 1; parathyroid hormone/parathyroid hormone-related peptide receptor; parathyroid hormone/parathyroid hormone-related protein receptor; PFE; PPR; PTH; PTH/PTHr receptor; PTH/PTHrP receptor; PTH/PTHrP type I receptor; PTH1 receptor; PTH1R; Pthr; Pthr1; PTHrel; PTH-related peptide receptor; PTHrP-R; PTHrP-receptor; seven transmembrane helix receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5745 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q03431 | |
Pth1r | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human PTH1R. | |
50 μg | |
Primary | |
Human, Non-human Primate | |
Antibody | |
IgG |