Learn More
RAB2A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521754
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: H1299, HeLaS3. Predicted reactivity: Dog (100%), Rabbit (100%), Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RAB2A Polyclonal antibody specifically detects RAB2A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
RAB2A | |
Polyclonal | |
Unconjugated | |
Rab2a | |
9330148M11Rik; C80220; GTP binding protein; GTP-binding protein; LHX; OTTMUSP00000004796; rab 2; RAB2; RAB2, member RAS oncogene family; Rab2a; RAB2A, member RAS oncogene family; ras-related protein Rab-2A; RP23-183L6.1; small GTP binding protein RAB2A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5862 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.51 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P61019 | |
Rab2a | |
Synthetic peptide corresponding to a region within amino acids 148 and 212 of RAB2A (Uniprot ID#P61019) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |