missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pan Ras Monoclonal Antibody (Ras10), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1012
Code : LD
Additional Details : Weight : 0.10000kg
Description
This antibody recognizes normal p21, activated p21, and p21 from each of the Harvey, Kirsten and N-ras families.
Pan Ras Monoclonal antibody specifically detects Pan Ras in Human, Mouse, Non-human Primate samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot.Specifications
Pan Ras | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
P01111, P01112, P01116, P08556, P32883, Q61411 | |
HRAS, KRAS, Nras | |
Purified recombinant Ras protein | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Ras10 | |
Unconjugated | |
HRAS | |
AI929937; ALPS4; AV095280; C-BAS/HAS; cellular c-Ki-ras2; cellular c-Ki-ras2 proto-oncogene; CFC2; c-Ha-ras; c-Ha-ras p21 protein; c-Ha-ras transgene; C-HA-RAS1; c-has/bas p21 protein; C-H-RAS; c-Ki-ras; c-Kirsten-ras protein; c-Kirsten-ras proto-oncogene; c-K-ras; c-K-ras2 protein; CMNS; c-rasHa; c-ras-Ki-2 activated oncogene; CTLO; etID18709.8; fa04e08; fc14b12; fc23g10; fj89d12; fj99a10; GTP- and GDP-binding peptide B; GTPase HRas; GTPase HRas, N-terminally processed; GTPase Hras1; GTPase KRas; GTPase KRas, N-terminally processed; GTPase NRas; HAMSV; Ha-ras; Ha-Ras1 proto-oncoprotein; Harvey ras1 protein; Harvey rat sarcoma viral (v-Ha-ras) oncogene homolog; Harvey rat sarcoma viral oncogene homolog; Harvey rat sarcoma viral oncoprotein; Harvey rat sarcoma virus oncogene; Harvey rat sarcoma virus oncogene 1; Harvey-ras; hras; H-ras; H-ras 1 protein; HRas proto-oncogene, GTPase; HRas proto-oncogene, GTPase a; HRAS1; H-Ras-1; Hras-1; hrasa; H-RASIDX; hypothetical protein LOC445289; hy | |
Mouse | |
Protein G | |
RUO | |
15461, 16653, 18176, 3265, 3845, 4893 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |