Learn More
RCC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA528531
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (99%), Zebrafish (91%), Xenopus laevis (88%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RCC2 Polyclonal antibody specifically detects RCC2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
RCC2 | |
Polyclonal | |
Unconjugated | |
RCC2 | |
2610510H01Rik; 2610529N02Rik; AA536646; AA675016; KIAA1470; mKIAA1470; protein RCC2; Protein RCC2 homolog; RCC1-like; RCC1-like protein TD-60; RCC2; regulator of chromosome condensation 2; RGD1309986; Td60; td-60; Telophase disk protein of 60 kDa; wu:fb36a03; zgc:77115 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
108911, 55920 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8BK67, Q9P258 | |
RCC2 | |
Recombinant fragment corresponding to a region within amino acids 250 and 522 of Human RCC2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |