Learn More
Heparin Cofactor II Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen PA143055
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-43055 detects SERPIND1 in human samples. PA1-43055 has successfully been used in Western blot and ELISA applications. PA1-43055 was derived from human heparin cofactor II.
Heparin Cofactor II Polyclonal antibody specifically detects Heparin Cofactor II in Human samples. It is validated for ELISA, Western Blot.Specifications
Heparin Cofactor II | |
Polyclonal | |
Unconjugated | |
Serpind1 | |
AA985900; AI303446; D22S673; HC2; Hcf2; HCII; HC-II; Heparin cofactor 2; heparin cofactor II; HLS2; leuserpin 2; leuserpin-2; LS2; Protease inhibitor leuserpin-2; rls2var1; serine (or cysteine) peptidase inhibitor, clade D, member 1; serine (or cysteine) proteinase inhibitor, clade D (heparin cofactor), member 1; serine (or cysteine) proteinase inhibitor, clade D, member 1; serpin D1; serpin family D member 1; serpin peptidase inhibitor, clade D (heparin cofactor), member 1; Serpind1; THPH10 | |
Sheep | |
Ammonium sulfate precipitation, Ion-exchange chromatography | |
RUO | |
3053 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
8.9 mg/mL | |
50% water with 50% glycerol and no preservative | |
P05546 | |
Serpind1 | |
Human Heparin Cofactor II. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |