missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A17 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA526180
Code : NEW
Additional Details : Weight : 0.01000kg
Description
SLC25A17 Polyclonal antibody specifically detects SLC25A17 in Human, Mouse samples. It is validated for Flow Cytometry, Western Blot.Specifications
SLC25A17 | |
Polyclonal | |
Unconjugated | |
SLC25A17 | |
34 kDa peroxisomal membrane protein; 34kDa; 47kDa; peroxisomal integral membrane protein 47; peroxisomal membrane protein PMP34; peroxisomal membrane protein, 34 kDa; Pmp34; Pmp35; Pmp47; SLC25A17; solute carrier family 25 (mitochondrial carrier, peroxisomal membrane protein), member 17; solute carrier family 25 (mitochondrial carrier; peroxisomal membrane protein, 34kDa), member 17; solute carrier family 25 member 17; solute carrier family 25, member 17 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10478, 20524 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.35 mg/mL | |
PBS with 0.09% sodium azide | |
O43808, O70579 | |
SLC25A17 | |
KLH conjugated synthetic peptide between 17-44 amino acids from the N-terminal region of human SLC25A17 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |