Learn More
TCR V beta 5a Monoclonal Antibody (1C1), FITC, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen TCR2642
340.01 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : A0
Additional Details : Weight : 0.01000kg
Description
TCR2642 targets TCR V beta 5a in FACS, IHC(F), and TCA applications and shows reactivity with Human, Mouse and Non-human primate samples. The TCR2642 immunogen is human TCR V beta 5(a).
TCR V beta 5a Monoclonal antibody specifically detects TCR V beta 5a in Human, Mouse, Non-human Primate samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen), T-Cell Activation.Specifications
TCR V beta 5a | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.5% BSA and 0.1% sodium azide | |
0 | |
TCRB | |
Human TCR V beta 5(a) | |
200 Tests | |
Primary | |
Human, Mouse, Non-human Primate | |
Antibody | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen), T-Cell Activation | |
1C1 | |
FITC | |
TCRB | |
T cell receptor beta chain; T cell receptor beta locus; TCRB; TCRbeta; Tib; TRB; TRB@ | |
Mouse | |
Protein G | |
RUO | |
21577 | |
4° C, store in dark | |
Liquid |