missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAF6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 380900
Additional Details : Weight : 0.50000kg
Description
TRAF6 Polyclonal antibody specifically detects TRAF6 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.Specifications
TRAF6 | |
Polyclonal | |
Unconjugated | |
TRAF6 | |
2310003F17Rik; AI851288; C630032O20Rik; E3 ubiquitin-protein ligase TRAF6; interleukin-1 signal transducer; MGC:3310; RING finger protein 85; RING-type E3 ubiquitin transferase TRAF6; RNF85; TNF receptor associated factor 6; TNF receptor-associated factor 6; TNF receptor-associated factor 6, E3 ubiquitin protein ligase; Traf6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
22034, 7189 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
P70196, Q9Y4K3 | |
TRAF6 | |
Raised against the C-term of human TRAF6. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |