Learn More
VWF Monoclonal Antibody (F8/86), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA514029
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-14029 targets von Willebrand Factor in IHC (P) applications and shows reactivity with Human samples. The MA5-14029 immunogen is partially purified von Willebrand factor from human placenta.
VWF Monoclonal antibody specifically detects VWF in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
VWF | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
P04275 | |
VWF | |
Partially purified von Willebrand factor from human placenta | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
F8/86 | |
Unconjugated | |
VWF | |
6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 | |
Mouse | |
Protein G | |
RUO | |
7450 | |
4° C | |
Liquid |