missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRPV1 Polyclonal Antibody, Invitrogen™
Guinea Pig Polyclonal Antibody
Brand: Invitrogen PA129770
Code : LE
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TRPV1 Polyclonal antibody specifically detects TRPV1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen).Specifications
TRPV1 | |
Polyclonal | |
Unconjugated | |
TRPV1 | |
Capsaicin receptor; cation channel; DKFZp434K0220; non-selective cation channel; osm-9-like TRP channel 1; OTRPC1; si:dkey-9e15.2; transient receptor potential cation channel subfamily V member 1; transient receptor potential cation channel V1; transient receptor potential cation channel, subfamily V, member 1; transient receptor potential V1; transient receptor potential V1 channel; transient receptor potential vanilloid 1 supraoptic nucleus; transient receptor potential vanilloid 1a; transient receptor potential vanilloid 1b; TRPV1; TRPV1/2; TRPV1_SON; TRPV1alpha; TRPV1beta; Vanilloid receptor; vanilloid receptor 1; vanilloid receptor subtype 1; vanilloid receptor type 1 like protein 1; Vanilloid receptor type 1-like; vanilloid type 1 receptor; VR.5'sv; Vr1; VR-1; Vr1l; Vr1l1 | |
Guinea Pig | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry (Frozen) | |
Conc. not determined | |
whole serum with 0.02% sodium azide | |
O35433, Q704Y3, Q8NER1 | |
TRPV1 | |
Peptide corresponding to residues YTGSLKPEDAEVFKDSMVPGEK of the carboxy-terminus of rat VR1 | |
RUO | |
193034, 7442, 83810 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |