missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530991
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
WDR4 Polyclonal antibody specifically detects WDR4 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
WDR4 | |
Polyclonal | |
Unconjugated | |
WDR4 | |
AI415180; AI448349; D530049K22Rik; hWH; mWH; Protein Wuho homolog; TRM82; TRM82 tRNA methyltransferase 82 homolog; TRMT82; tRNA (guanine-N(7)-)-methyltransferase non-catalytic subunit WDR4; tRNA (guanine-N(7)-)-methyltransferase subunit WDR4; WD repeat domain 4; WD repeat-containing protein 4; WD40 repeat protein 4; WDR4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10785 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.71 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P57081 | |
WDR4 | |
Recombinant protein fragment corresponding to a region within amino acids 98 and 365 of Human WDR4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |