missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF154 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543100
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QRCLYRDVML ENLALLTSLD VHHQKQHLGE KHFRSNVGRA LFVKTCTFHV Sequence homology: Horse: 85%; Human: 100%; Pig: 77%
ZNF154 Polyclonal antibody specifically detects ZNF154 in Human samples. It is validated for Western Blot.Specifications
ZNF154 | |
Polyclonal | |
Unconjugated | |
ZNF154 | |
KIAA2003; pHZ-92; Zinc finger protein 154; ZNF154 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7710 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13106 | |
ZNF154 | |
Synthetic peptide directed towards the N-terminal of human ZNF154 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |