missing translation for 'onlineSavingsMsg'
Learn More

Antimony, AAS standard solution, Specpure™ Sb 1000μg/mL, Thermo Scientific Chemicals

Catalog Number p-7082691
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
7732-18-5
Sb in 30% HCl
MFCD00134030
This item is not returnable. View return policy
7732-18-5
Sb in 30% HCl
MFCD00134030

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Concentration Sb 1000mug/mL
Quantity 100 mL
UN Number UN1789
Odor Acidic
Physical Form Liquid
Chemical Name or Material Antimony, AAS standard solution, Specpure™

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'