missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Antimony(III) oxide, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
2500 g
Description
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
Antimony(III) oxide is used as a catalyst, vulcanization of rubber, an opacifying agent for glasses, ceramics and enamels. It is used as a flame retardant.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
Specifications
Boiling Point | 1550°C (sublimation) |
Melting Point | 656°C |
Quantity | 500 g |
Merck Index | 14,711 |
Solubility Information | Insoluble in water. |
Formula Weight | 291.5 |
Percent Purity | 99% |
Odor | Odorless |
Density | 5.67 |
Chemical Name or Material | Antimony (III) oxide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'