missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ APET Liners for Safety Trays
Environmental friendly, easy to clean and maintain for routine work and storage in safe areas.
71.60 € - 186.00 €
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
12307207
|
Fisherbrand™
|
260 x 460mm |
71.60 €
Pack of 25 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||||
12317207
|
Fisherbrand™
|
340 x 540mm |
73.10 €
Pack of 25 |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
12327207
|
Fisherbrand™
|
540 x 570mm |
86.40 €
Pack of 25 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||||
12337207
|
Fisherbrand™
|
540 x 680mm |
118.00 €
Pack of 25 |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
12347207
|
Fisherbrand™
|
460 x 700mm |
186.00 €
Pack of 25 |
In Stock
Log in to see stock. |
|||||
Description
- Clears up spillages easily
- Environmental friendly
Specifications
APET | |
Liners | |
25 | |
Rectangular |