missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apoptosis Activator 2, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 mg
25 mg
Description
Activates apoptosis in cells; shows selectivity for cancer cells against normal cell lines
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Description | Orange |
Quantity | 25 mg |
Formula Weight | 306.14g/mol |
Sensitivity | Light sensitive |
Product Type | Apoptosis Reagent |
Content And Storage | 2°C to 8°C |
For Use With (Application) | Activates apoptosis in cells, shows selectivity for cancer cells against normal cell lines |
InChI Key | KGRJPLRFGLMQMV-UHFFFAOYSA-N |
SMILES | ClC1=CC=C(CN2C(=O)C(=O)C3=CC=CC=C23)C=C1Cl |
IUPAC Name | 1-[(3,4-dichlorophenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'