Learn More
Karl Fischer Aqualine™ Electrolyte AG, for Karl Fischer Titration By Coulometry, Fisher Chemical™
Nonhalogenated anolyte solution for general use in conventional cells with a diaphragm
Brand: Fisher Chemical K/2520/08
133.00 EUR valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Code : 24
Additional Details : CAS Number : 67-56-1 Weight : 0.87000kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Description
Nonhalogenated anolyte solution for general use in conventional cells with a diaphragm
- Anolyte solution for use in conventional cells with a diaphragm
- Source of electrolyte required for anodic oxidation for coulometric analysis
- Contains methanol as a solvent
Specifications
Colorless | |
Aqualine™ Electrolyte AG | |
108-75-8,111-42-2,7446-09-5,7553-56-2 | |
Liquid | |
3097387 |
500 mL | |
Karl Fischer Reagent | |
Amber glass bottle | |
Pass Test |