missing translation for 'onlineSavingsMsg'
Learn More

Arecoline hydrobromide, 98+%, ACROS Organics™

Brand:  Acros Organics 250131000

Code : 29

Additional Details : CAS Number : 300-08-3 Weight : 0.15000kg Transport : UN number : 1544 Chem class : 6.1 Pack group : III


Packaging Glass bottle
Quantity 100g

 View more versions of this product

Product Code. 10023813

  • 287.00 € / 100g

Estimated Shipment
24-05-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

300-08-3
236.109
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
9301
CN1CCC=C(C1)C(=O)OC.Br
C8H14BrNO2
MFCD00039041
arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide

Specifications

Arecoline hydrobromide
Glass bottle
168°C to 174°C
300-08-3
C8H14BrNO2
15, 768
AXOJRQLKMVSHHZ-UHFFFAOYSA-N
methyl 1-methyl-3,6-dihydro-2H-pyridine-5-carboxylate;hydrobromide
9301
Powder
Authentic
White to Yellow
100g
98+%
MFCD00039041
arecoline hydrobromide, arecoline bromide, taeniolin, arecoline hbr, methyl 1-methyl-1,2,5,6-tetrahydropyridine-3-carboxylate hydrobromide, arekolinhydrobromid german, unii-24s79b9cx7, methyl 1,2,5,6-tetrahydro-1-methylnicotinate hydrobromide, n-methyltetrahydronicotinic acid methyl ester hydrobromide
CN1CCC=C(C1)C(=O)OC.Br
236.109
236.11
≥98.0%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Arecoline hydrobromide

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3

Hazard Statement
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS Documents