Learn More
Thermo Scientific™ Armadillo PCR Plate, 384-well, yellow, clear wells, barcoded
Combine the rigidity of a polycarbonate frame with thin walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.
Brand:  Thermo Scientific™ BC2384Y
Code : LD
Additional Details : Weight : 1.00000kg
Product Code. 12607446
  • 408.00 € / Pack of 50
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Armadillo PCR plates combine the rigidity of a polycarbonate frame with thin walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.
Thermo Scientific Armadillo PCR plates combine the rigidity of a polycarbonate frame with thin-walled polypropylene wells to provide superior thermal cycling performance under all conditions without warping.
They are available in 96- and 384-well formats, in multiple colors, and can be ordered with a standard 128 barcode or custom barcoding. All formats are available with clear or white wells; choose white for superior qPCR results. The specially-designed warp resistant frame and multiple format options make Armadillo PCR plates the ultimate choice for high-throughput PCR and automated handling.
  • Polycarbonate frame for warp resistant thermal cycling
  • Enhanced mechanical stability for robotic handling
  • Thin walled polypropylene wells for optimal heat transfer
  • Well shape optimized for maximum sample recovery
  • Flat alpha-numeric lettering and raised rim well design for improved heat sealing
  • Optically clear deck allows for easy visibility of wells
  • Multiple frame color options, all available in both clear (for PCR) and white colored wells (for qPCR)
  • Maximum well volume 40μL
  • Maximum working volume 25μL
Specifications
Specifications
Includes Barcode
50 Plate
Yellow, Clear
Real-Time PCR Systems or Thermal Cyclers
Polycarbonate, Polypropylene
384
Standard
Certified RNase/DNase-free
Armadillo
PCR Microplate
Skirted
40 μL
25 μL
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
This product is sold under license from Eppendorf AG.