Learn More
Thermo Scientific™ ART™ Non-Filtered Standard Hinged Rack Pipette Tips
Access tips one-handed with this versatile hinged rack, which is designed for the way you work.
74.90 € - 121.00 €
Specifications
Filtered | Non-Filtered |
---|---|
Color | Natural |
Model | ART Non-filtered |
Clean Claims | DNase, RNase, DNA, ATP and Endotoxin Free |
Format | Hinged Rack |
Product Code | Brand | Description | Volume (Metric) | Sterility | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Sterility | Fit for Purpose | Price | Quantity & Availability | ||
15163626
|
Thermo Scientific™
3101-05-HR |
ART Non-filtered, LR, Sterile, Hinged Rack | 1000 μL | Sterile | Sterile |
107.00 €
Pack of 768 |
In Stock
Log in to see stock. |
||
15153626
|
Thermo Scientific™
3102-05-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 1000 μL | Non-sterile | Certified Quality |
94.95 €
Pack of 768 |
In Stock
Log in to see stock. |
||
15133626
|
Thermo Scientific™
3101-HR |
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack | 1000 μL | Sterile | Sterile |
119.00 €
Pack of 768 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15143626
|
Thermo Scientific™
3102-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 1000 μL | Non-sterile | Certified Quality |
77.55 €
Pack of 768 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15143616
|
Thermo Scientific™
3771-05-HR |
ART Non-filtered, LR, Sterile, Hinged Rack | 300 μL | Sterile | Sterile |
115.00 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15133616
|
Thermo Scientific™
3772-05-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 300 μL | Non-sterile | Certified Quality |
104.00 €
Pack of 960 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||
15113616
|
Thermo Scientific™
3772-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 300 μL | Non-sterile | Certified Quality |
94.40 €
Pack of 960 |
Estimated Shipment: 16-10-2024
Log in to see stock. |
||
15123616
|
Thermo Scientific™
3771-HR |
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack | 300 μL | Sterile | Sterile |
111.00 €
Pack of 960 |
In Stock
Log in to see stock. |
||
15153596
|
Thermo Scientific™
3551-05-HR |
ART Non-filtered, LR, Sterile, Hinged Rack | 200 μL | Sterile | Sterile |
98.25 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15133596
|
Thermo Scientific™
3552-05-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 200 μL | Non-sterile | Certified Quality |
84.75 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15123596
|
Thermo Scientific™
3551-HR |
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack | 200 μL | Sterile | Sterile |
121.00 €
Pack of 960 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||
15113596
|
Thermo Scientific™
3552-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 200 μL | Non-sterile | Certified Quality |
75.10 €
Pack of 960 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||
15133586
|
Thermo Scientific™
3521-05-HR |
ART Non-filtered, LR, Sterile, Hinged Rack | 20 μL | Sterile | Sterile |
98.25 €
Pack of 960 |
In Stock
Log in to see stock. |
||
15113586
|
Thermo Scientific™
3522-05-HR |
ART Non-filtered, LR, Non-sterile, Hinged Rack | 20 μL | Non-sterile | Certified Quality |
85.15 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15103586
|
Thermo Scientific™
3522-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 20 μL | Non-sterile | Certified Quality |
74.90 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15143576
|
Thermo Scientific™
3511-HR |
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack | 10 μL | Sterile | Sterile |
92.40 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15103576
|
Thermo Scientific™
3501-05-HR |
ART Non-filtered, LR, Sterile, Hinged Rack | 10 μL | Sterile | Sterile |
98.25 €
Pack of 960 |
In Stock
Log in to see stock. |
||
15143566
|
Thermo Scientific™
3502-05-HR |
ART Non-filtered, LR, Non-sterile, Hinged Rack | 10 μL | Non-sterile | Certified Quality |
82.35 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
15153566
|
Thermo Scientific™
3501-HR |
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack | 10 μL | Sterile | Sterile |
91.20 €
Pack of 960 |
Estimated Shipment: 04-09-2024
Log in to see stock. |
||
15123566
|
Thermo Scientific™
3502-HR |
ART Non-filtered, Non-Filtered, Non-sterile, Hinged Rack | 10 μL | Non-sterile | Certified Quality |
95.75 €
Pack of 960 |
In Stock
Log in to see stock. |
||
15183576
|
Thermo Scientific™
3521-HR |
ART Non-filtered, Non-Filtered, Sterile, Hinged Rack | 20 μL | Sterile | Sterile |
95.05 €
Pack of 960 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
Description
Access tips with one hand using Thermo ScientificTM ARTTM Nonfiltered Hinged Rack Pipette Tips, which is designed for the way you work. No longer worry about your tips spilling on your bench or floor. Maximize your productivity and easily transition from hinged to lift-off in just three easy steps. Simply unhinge, turn and realign the cover--now you have a traditional lift-off when you need it.
Features:
- Complete selection of non-filtered and specialty tips deliver proven ART quality, performance and reliability, backed by the highest Thermo Scientific standards and worldwide support
- Sterile tips are DNase, RNase, DNA, ATP, and Endotoxin Free
- Racks offer quick and easy access to pipette tips, saving valuable time
- Front latch closes and locks, ensuring safe and secure storage throughout
Specifications
Non-Filtered | |
ART Non-filtered | |
Hinged Rack |
Natural | |
DNase, RNase, DNA, ATP and Endotoxin Free | |
ART |