Learn More
Thermo Scientific™ ART™ Non-filtered Low Retention Pipette Tips in Lift-off Lid Rack
Specifically designed for eliminating sample retention within the pipette tip.
53.35 € - 86.60 €
Product Code | Brand | Description | Volume (Metric) | Tip Style | Sterility | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Volume (Metric) | Tip Style | Sterility | Price | Quantity & Availability | ||
10110062
|
Thermo Scientific™
3771-05 |
ART 300, Non-Filtered, Sterile, Rack | 300 μL | MicroPoint | Sterile |
86.60 €
Pack of 960 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||
10550682
|
Thermo Scientific™
3772-05 |
ART 300, Non-Filtered, Non-sterile, Rack | 300 μL | MicroPoint | Non-sterile |
78.00 €
Pack of 960 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||
10261722
|
Thermo Scientific™
3511-05 |
ART 10 REACH, Non-filtered, Sterile, Racked, Lift-off Lid | 10 μL | Low Retention | Sterile |
70.85 €
Pack of 960 |
|
||
10251722
|
Thermo Scientific™
3512-05 |
ART 10 REACH, Non-filtered, Non-sterile, Racked, Lift-off Lid | 10 μL | Low Retention | Non-sterile |
56.40 €
Pack of 960 |
In Stock
Log in to see stock. |
||
10241772
|
Thermo Scientific™
3521-05 |
ART 20, Low Retention, Non-filtered, Sterile, Racked, Lift-off Lid | 20 μL | Low Retention | Sterile |
73.50 €
Pack of 960 |
Estimated Shipment: 02-09-2024
Log in to see stock. |
||
10048481
|
Thermo Scientific™
3522-05 |
ART 20 Low Retention, Non-filtered, Non-sterile, Racked, Lift-off Lid | 20 μL | Low Retention | Non-sterile |
53.35 €
Pack of 960 |
Estimated Shipment: 16-09-2024
Log in to see stock. |
||
Description
Thermo Scientific™ ART™ Non-filtered Low Retention Pipette Tips in Lift-off Lid Rack are specifically designed for eliminating sample retention within the pipette tip.
- Inner surface is extremely hydrophobic, increasing sample reproducibility and sample accuracy
- Crystal clear tips allow you to easily see transparent samples
- Tips seal when exposed to potential contaminants
- DNase, RNase, DNA, ATP, and Endotoxin Free
Specifications
Non-Filtered | |
Clear | |
ART |