Learn More
Karl Hecht™ Assistent™ Filter Paper Circles
Provides for fast filtration in the laboratory. Karl Hecht™ Assistent™ Filter Paper Circles are grained and have a thickness measuring 0.20mm.
POA
Description
- Rate of filtration: 10 - 12s
- Available in a range of diameters
- Supplied in packs of 100
Specifications
Filter Paper Disc |